Round Plastic Bottles
Links
Save Now - Web Exclusive Deals
Take advantage of our web exclusive prices and place your order today.
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Bottles with Closure
Thermo Scientific™ Nalgene™ Heavy-Duty PPCO Vacuum Bottles with Closure: Lab Pack
Must Have
Complete Your Order
Complete your round plastic bottles order with these supplies.
  • Great deals
    Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
    Give new function to your plastic bottles with these replacement caps.
    Learn More
  • Great deals
    Thermo Scientific™ Nalgene™ Filling/Venting Closures
    Aseptic and leakproof handling solutions - compatible with Nalgene™ bottles and carboys.
    Learn More
Guides to Help You Choose
Thermo Scientific™ Nalgene™ Bottle and Carboy Technical Guide
Thermo Scientific™ Nalgene™ Bottle and Carboy Technical Guide
Get familiar with Nalgene™ products, certifications, and care instructions. The guide also includes a section about plastic resins and their compatibility with chemicals.
Download
Focus on Glassware & Plasticware
Focus on Glassware & Plasticware
Your essential guide to everyday labware. Resolve your doubts about different material types and learn how to care for your items. The guide also includes a chemical compatibility chart and an FAQ section.
Download
Shop Your Brands
Frequently Asked Questions
Can plastic bottles be autoclaved?
Generally, polypropylene, PTFE, PFA, and PMP materials can be autoclaved; however, temperatures higher than 80°C can affect the accuracy of volumetric vessels. Always refer back to the product technical guide and remember to loosen any caps before autoclaving.
What kind of detergents can I use on my plastic bottles?
Ideally, a low or non-alkaline detergent should be used as some plastics such as polystyrene and polycarbonate are susceptible to alkalis. Avoid abrasive materials and wash below 60°C if using an electrical washer. For further information check the product guide.
Which plastic materials have a good chemical compatibility?
Plastic bottles made of polypropylene (PPCO) and polyethylene (HDPE and LDPE) have good chemical compatibility, and bottles made of fluoropolymers (FEP/PFA) are even chemical-resistant and inert. Polycarbonate (PC) and polyethylene terephthalate glycol (PETG) bottles have lower chemical compatibility but offer higher visibility.
Browse Other Product Categories